Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Brand: Quickfit EX13/12
This item is not returnable.
View return policy
Description
- Designed to support Raschig rings within plain fractionating columns
- Support is passed through socket of column and rests at the constriction above the cone
- Manufactured from chemically resistant borosilicate glass