Learn More
Quickfit™ Borosilicate Glass Column Packing Support
Product Code. 11391875
Change view
Click to view available options
Material:
Pyrex Borosilicate Glass
Unit Size:
Each
Product Code. Material unitSize
11391875 Pyrex Borosilicate Glass Each
1 options
Product Code. 11391875
Brand: Quickfit EX13/23
Designed to support Raschig rings within plain fractionating columns
Designed to support Raschig rings within plain fractionating columns
  • Designed to support Raschig rings within plain fractionating columns
  • Support is passed through socket of column and rests at the constriction above the cone
  • Manufactured from chemically resistant borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Designed to support Raschig rings within plain fractionating columns
Type Column packing support
Material Pyrex Borosilicate Glass
For Use With (Equipment) Column