Learn More
Quickfit™ 20 mm Plastic Cap
Product Code. 15748027
Click to view available options
Thread Size (Metric):
L/ mm
Unit Size:
Pack of 5
Product Code. 15748027
Brand: Quickfit ML/CAP20
Only null left
Add to basket
Add to basket
Quickfit™ 20 mm Plastic Cap is a spare part for use with the ML1 microscale kit.
  • Spare screw cap for ML1 microscale kit and microscale dropping funnel
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Thread Size (Metric) L/ mm
Neck Finish L/ mm
Product Type Screw Cap
For Use With (Equipment) ML1 microscale kit, thermometer adapter
Closure Size 20 mm
Diameter (Metric) 20 mm