Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Product Code. 10100161 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
DN 100
DN 120
DN 150
DN 200
DN 60
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
10100161 DN 100 Each
10464691 DN 60 Each
10683331 DN 120 Each
12656203 DN 150 Each
10170201 DN 200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10100161 Supplier DWK Life Sciences™ Supplier No. 290714609
Only null left
Add to basket
Add to basket
The quick release clamp is used to close flat flange vessels.
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
  • With retaining clip
  • Quick closing with quick-release fastener
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Material Stainless Steel
For Use With (Equipment) Quick Release Clamp for Closure
For Use With (Application) Quick release clamp for closure; Accessories for flat flange vessels
Size DN 100