Learn More
Epredia™ Quick Release Clamp, Aligner
Fix cassettes or embedding rings both longitudinally and horizontally, with orientation on X-, Y-, and Z-axes using the Epredia™ Quick Release Clamp.
Description
Description
Universal Cassette Clamps
  • For all types of commercially available cassettes
  • Silver model is for HM 325, HM 355S, and HM 340E models with serial numbers above 34000
Quick Release Clamp
  • For embedding rings
Zero Orientation Device
  • Positions specimen holder flat against the knife or blade (aligns X, Y axis)