Learn More
SCAT Quick-Lock Connectors
Product Code. 17342840 Shop All SCAT Europe Products
Change view
Click to view available options
Product Type:
Quick-Lock Connector (f)
Quick-Lock Connector (m) for 1 Capillary Tube
Quick-Lock Connector (m) for 1 Safety Cap
Quick-Lock Connector Set I
Quick-Lock Connector Set II
Unit Size:
1 set
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17342840 Quick-Lock Connector (m) for 1 Capillary Tube Each
17322840 Quick-Lock Connector (f) Each
17332840 Quick-Lock Connector (m) for 1 Safety Cap Each
17312860 Quick-Lock Connector Set I 1 set
17322860 Quick-Lock Connector Set II 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17342840 Supplier SCAT Supplier No. 360190
  • ONLINE EXCLUSIVE PRICE
    £33.80
    £30.59 / Pack of 1
    Save £3.21
    9% Off
View all promotions
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Quick-Lock Connectors provide easy and safe connection of capillary tubing and SCAT Safety Caps. Connect and disconnect with a click - without dripping or air bubbles. The valve closes automatically when disconnected. Included PFA fittings allow all common hose diameters to be connected.
The Quick-Lock Connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
  • Made of chemically resistant glass-filled polypropylene, PEEK™ materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1.6 (1/16 in.) / 2.3 /3.2 (1/8 in.) mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1.0 bar pressure resistance
Specifications
For Use With (Equipment) Capillaries, Tubes and Safety Caps
Includes 1 x PFA-fitting 1.6/2.3/3.2 mm ID
For Use With (Application) HPLC Systems
Product Type Quick-Lock Connector (m) for 1 Capillary Tube