missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Quick Charger
Click to view available options
Color:
Black
Unit Size:
Each
Description
- Accessory for BMP71 printer.
Specifications
Specifications
| Product Type | AC Adapter/Battery Charger |
| Color | Black |
| For Use With (Equipment) | BMP71 Label Printer |
| Description | Quick Charger, US 120 V |
| Format | Charger |
| Quantity | 1 Ea. |
| Voltage | 120 V |
| Alternate Part Number | M71-QC-US |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction