Learn More
Eppendorf™ Quick-change Platform Kit for Innova™ 2300 Shaker
Product Code. 11668633
Click to view available options
For Use With (Equipment):
Innova™ 2300 Shaker
Unit Size:
Each
Product Code. 11668633
Brand: Eppendorf™ M11919904
This quick-change platform kit enables the user to rapidly attach platforms in place to the model 2300 shaker without the need for tools.
  • Quick-change kit
  • No tools required
  • Platforms sold separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Platform Attachment without Tools
For Use With (Equipment) Innova™ 2300 Shaker