missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine N-Oxide, TRC

Product Code. 30006632
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006632 5 mg 5mg
30008631 1 mg 1mg
30007682 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006632 Supplier Toronto Research Chemicals Supplier No. TRCQ5100205MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076199-40-0
Molecular Formula C21 H25 N3 O3 S
Molecular Weight (g/mol) 399.51
Synonym 2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)-1-oxidopiperazin-1-yl]ethoxy]ethanol, Quetiapine Fumarate Imp. H (EP), Quetiapine N-Oxide
IUPAC Name 2-[2-(4-benzo[b][1,4]benzothiazepin-6-yl-1-oxidopiperazin-1-ium-1-yl)ethoxy]ethanol
SMILES OCCOCC[N+]1([O-])CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H25N3O3S/c25-14-16-27-15-13-24(26)11-9-23(10-12-24)21-17-5-1-3-7-19(17)28-20-8-4-2-6-18(20)22-21/h1-8,25H,9-16H2
Formula Weight 399.1617
Percent Purity >95%
Chemical Name or Material 2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)-1-oxidopiperazin-1-yl]ethoxy]ethanol (Quetiapine N-Oxide)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.