missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine, MedChemExpress

Product Code. p-200099393
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16476088

Brand: Medchem Express HY14544

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quetiapine (ICI204636) is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Quetiapine (ICI204636) is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Specifications

Chemical Name or Material Quetiapine
CAS 111974-69-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H25N3O2S
Quantity 10 mg
Synonym ICI204636
Solubility Information Ethanol : 100 mg/mL (260.75 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (260.75 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.26 mM; ultrasonic and warming and heat to 50°C)
SMILES OCCOCCN(CC1)CCN1C2=NC3=CC=CC=C3SC4=C2C=CC=C4
Molecular Weight (g/mol) 383.51
Formula Weight 383.51
Percent Purity 99.96%
Grade Research
Physical Form Oil
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.