missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quetiapine, MedChemExpress
Description
Quetiapine (ICI204636) is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.
Specifications
Specifications
| Chemical Name or Material | Quetiapine |
| CAS | 111974-69-7 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H25N3O2S |
| Quantity | 10 mg |
| Synonym | ICI204636 |
| Solubility Information | Ethanol : 100 mg/mL (260.75 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (260.75 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.26 mM; ultrasonic and warming and heat to 50°C) |
| SMILES | OCCOCCN(CC1)CCN1C2=NC3=CC=CC=C3SC4=C2C=CC=C4 |
| Molecular Weight (g/mol) | 383.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?