missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Hemifumarate, TRC

Product Code. 17904305
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904305 500 mg 500mg
17973305 100 mg 100mg
17971715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904305 Supplier Toronto Research Chemicals Supplier No. TRCQ510000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111974-72-2
Molecular Formula 2 C21 H25 N3 O2 S . C4 H4 O4
Molecular Weight (g/mol) 883.09
Synonym Quetiapine Fumarate, Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2E)-2-butenedioate (2:1), Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (2E)-2-butenedioate (2:1) (salt) (9CI), Ethanol, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-, (E)-2-butenedioate (2:1) (salt), Dibenzo[b,f][1,4]thiazepine, ethanol deriv., ICI 204636, Quetiapine fumarate, Quetiapine hemifumarate, Seroquel, ZD 5077, ZM 204636
IUPAC Name 2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethanol;(E)-but-2-enedioic acid
SMILES OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24.OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68.OC(=O)\C=C\C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C21H25N3O2S.C4H4O4/c2*25-14-16-26-15-13-23-9-11-24(12-10-23)21-17-5-1-3-7-19(17)27-20-8-4-2-6-18(20)22-21;5-3(6)1-2-4(7)8/h2*1-8,25H,9-16H2;1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formula Weight 882.3445
Percent Purity >95%
Chemical Name or Material Quetiapine fumarate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.