missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine hemifumarate, MedChemExpress

Product Code. 16400768
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16400768

Brand: Medchem Express HYB0031

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quetiapine hemifumarate is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine hemifumarate is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine hemifumarate has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Quetiapine hemifumarate is a 5-HT receptors agonist with a pEC50 of 4.77 for human 5-HT1A receptor. Quetiapine hemifumarate is a dopamine receptor antagonist with a pIC50 of 6.33 for human D2 receptor. Quetiapine hemifumarate has moderate to high affinity for the human D2, HT1A, 5-HT2A, 5-HT2C receptor with pKis of 7.25, 5.74, 7.54, 5.55. Antidepressant and anxiolytic effects.

Specifications

Chemical Name or Material Quetiapine hemifumarate
CAS 111974-72-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27N3O4S
Quantity 5 g
Solubility Information DMSO : 50 mg/mL (113.24 mM; Need ultrasonic) ∣H2O : 1.25 mg/mL (2.83 mM; Need ultrasonic)
SMILES OCCOCCN(CC1)CCN1C2=NC(C=CC=C3)=C3SC4=C2C=CC=C4.O=C(O)/C=C/C(O)=O.[0.5]
Molecular Weight (g/mol) 441.54
Formula Weight 441.54
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.