missing translation for 'onlineSavingsMsg'
Learn More

Quetiapine Carboxylate Impurity, TRC

Product Code. 30418663
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418663 25 mg 25mg
30413411 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30418663 Supplier Toronto Research Chemicals Supplier No. TRCQ50998025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011758-00-1
Molecular Formula C22 H25 N3 O4 S
Molecular Weight (g/mol) 427.52
Synonym 1-Piperazinecarboxylic acid, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, 2-(2-hydroxyethoxy)ethyl ester, 2-(2-Hydroxyethoxy)ethyl 4-Dibenzo[b,f][1,4]thiazepin-11-ylpiperazine-1-carboxylate
IUPAC Name 2-(2-hydroxyethoxy)ethyl 4-benzo[b][1,4]benzothiazepin-6-ylpiperazine-1-carboxylate
SMILES OCCOCCOC(=O)N1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H25N3O4S/c26-13-14-28-15-16-29-22(27)25-11-9-24(10-12-25)21-17-5-1-3-7-19(17)30-20-8-4-2-6-18(20)23-21/h1-8,26H,9-16H2
Formula Weight 427.1566
Chemical Name or Material 2-(2-Hydroxyethoxy)ethyl 4-Dibenzo[b,f][1,4]thiazepin-11-ylpiperazine-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.