Learn More
Specac™ Quest ATR Spectroscopy ZnSe Puck
Product Code. 15230128
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Quest Single Reflection ATR Accessory GS10800-X
Product Code. For Use With (Equipment)
15230128 Quest Single Reflection ATR Accessory GS10800-X
1 options
Product Code. 15230128 Supplier Specac™ Supplier No. GS10812
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
ZnSe Puck for use in the Quest single reflection ATR Accessory GS10800-X. Ideal for softer materials.
Specifications
For Use With (Equipment) Quest Single Reflection ATR Accessory GS10800-X