Learn More
Specac™ Quest ATR Spectroscopy Ge Crystal Puck
Product Code. 15240128
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Quest Single Reflection ATR Accessory GS10800-X
Product Code. 15240128
Brand: Specac™ GS10813
Ge Puck for use in the Quest single reflection ATR Accessory GS10800-X. Ideal for strongly absorbing samples.
Specifications
For Use With (Equipment) Quest Single Reflection ATR Accessory GS10800-X