Learn More
Specac™ Quest ATR Spectroscopy Diamond Extended Range Puck
Product Code. 15220128
Encompass_Preferred
Click to view available options
For Use With (Equipment):
Quest Single Reflection ATR Accessory GS10800-X
Product Code. 15220128
Brand: Specac™ GS10811
Extended Range Diamond Crystal Puck for use in the Quest single reflection ATR Accessory GS10800-X. This Puck contains a strong and durable monolithic diamond. Designed for use in the mid- and far- infrared (10000 to 40).
Specifications
For Use With (Equipment) Quest Single Reflection ATR Accessory GS10800-X