missing translation for 'onlineSavingsMsg'
Learn More

Quercitrin, MedChemExpress

Product Code. 16449076
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449076

Brand: Medchem Express HYN0418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Quercitrin is a natural compound found in Tartary buckwheat with a potential anti-inflammation effect that is used to treat heart and vascular conditions.

Quercitrin is a natural compound found in Tartary buckwheat with a potential anti-inflammation effect that is used to treat heart and vascular conditions.

Specifications

Chemical Name or Material Quercitrin
CAS 522-12-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20O11
Quantity 100 mg
Synonym Quercetin 3-rhamnoside
Solubility Information DMSO : 125 mg/mL (278.78 mM; Need ultrasonic)
SMILES OC1=CC(O)=C(C(C(O[C@H]2[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O2)=C(C3=CC(O)=C(O)C=C3)O4)=O)C4=C1
Molecular Weight (g/mol) 448.38
Formula Weight 448.38
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.