missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Quercetin (Hydrate)

Product Code. 17180292
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17180292 10 g 10g
17170292 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17180292 Supplier STEMCELL Technologies Supplier No. 73934
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

mTOR, PI3K/AKT, NF-kB, and tyrosine kinase pathway inhibitor; Inhibits PI3K and SRC kinases

  • Quercetin inhibits phosphoinositide 3-kinase isoforms (beta: IC50 = 5.4 μM; gamma: IC50 = 2.4 μM; delta: IC50 = 3.0 μM) and SRC kinases but only shows slight inhibitory activity against serine/threonine kinases ERK1/2 and p38 alpha, as well as protein kinase C (Navarro-Nunez et al.)
  • Stimulates glycolytic metabolism and promotes reprogramming of somatic cells (Zhang et al.)
  • Enhances reprogramming of human primary somatic cells to induced pluripotent stem cells (Zhu et al.)
  • Induces hypoxia-inducible factor-1α (HIF-1α) and inhibits cell proliferation (Triantafyllou et al.)
  • Promotes apoptosis of glioblastoma cells when used together with temozolomide (Sang et al.)
  • Reduces tumorigenicity of pancreatic cancer cells (Fan et al.)

Specifications

CAS 849061-97-8
Quantity 10 g
Molecular Formula C15H10O7 · XH2O
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.