Learn More
Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits
Use Thermo Scientific™ QuEChERS Extraction Kits for the EN 15662 Method to analyze complex sample matrices for residues, toxins, and other contaminants.
£124.08 - £161.31
Specifications
Type | QuEChERS Extraction Kit |
---|---|
Method | QuEChERS sample preparation for EN 15662 Method |
Sample Size | 10 g sample |
Product Code | Brand | Description | Includes | Salt Composition | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Salt Composition | Price | Quantity & Availability | |||
17369822
|
Thermo Scientific™
S1-10-EN-CH-KIT |
QuEChERS EN 15662 Method Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl, 0.5 g Na-Citrate Sesquihydrate, 1 g Na-Citrate in Pouches; Ceramic Homogenizers; Empty 50 mL Tubes; 50/Pk. | Salts in Pouches, Tubes, Ceramic Homogenisers | 4 g MgSO4, 1 g NaCl, 0.5 g Na-Citrate Sesquihydrate, 1 g Na-Citrate |
£161.31
Pack of 50 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
17379822
|
Thermo Scientific™
S1-10-EN-KIT |
QuEChERS EN 15662 Method Extraction Kit for 10 g sample; 4 g Anhydrous MgSO4, 0.5 g Disodium Citrate, 1 g Trisodium Citrate, 1 g NaCl in Pouches; Empty 50 mL Tubes; 50/Pk. | Salts in Pouches, Tubes | 4 g Anhydrous MgSO4, 0.5 g Disodium Citrate, 1 g Trisodium Citrate, 1 g NaCl |
£144.76
Pack of 50 |
In Stock
Log in to see stock. |
|||
17389822
|
Thermo Scientific™
S1-10-EN-POT |
QuEChERS EN 15662 Method Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl, 1 g Na-Citrate, 0.5 g Disodium Citrate Sesquihydrate in Pouches; No Tubes; 50/Pk. | Salts in Pouches | 4 g MgSO4, 1 g NaCl, 1 g Na-Citrate, 0.5 g Disodium Citrate Sesquihydrate |
£124.08
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis of pesticide residues, veterinary drug residues, and toxins using the EN 15662 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.
Thermo Scientific™ QuEChERS EN 15662 Method Extraction Kits make sample preparation straight-forward and efficient, enabling you to confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.
Three different extraction kit formats are available for 10 g samples.
- Extraction Salts in Pouches, no Tubes
- Extraction Salts in Pouches with 50 mL Tubes
- Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers
What is the purpose of the chemicals used in the EN 15662 Method?
- Acetonitrile: used to perform single-phase extraction on a sample
- Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer; removes remaining water from sample
- NaCl: induces phase separation between water content in sample and acetonitrile layer; removes residual water from sample
- Trisodium Citrate: buffers the sample to stabilize pH
- Disodium Citrate Sesquihydrate: buffers the sample to stabilize pH
- PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample
- C18: removes fats, sterols, and other non-polar interferences from sample
- GCB: (graphitized carbon black) removes pigment (not recommended for use with planar pesticides)
Specifications
QuEChERS Extraction Kit | |
QuEChERS sample preparation for EN 15662 Method | |
10 g sample |