Learn More
Thermo Scientific™ QuEChERS 500 mg Silica Sorbent with C18 (endcapped) in 50 mL Centrifuge Tubes
Shop All Thermo Scientific ProductsDescription
Thermo Scientific QuEChERS Silica Sorbent with C18 (endcapped) in Centrifuge Tubes are designed for efficient sample cleanup in QuEChERS extractions. The pre-measured sorbent ensures consistent performance, while the endcapped C18 provides enhanced retention of non-polar compounds. Packaged in convenient centrifuge tubes, these sorbents streamline your laboratory workflow.
- Silica sorbent with C18 (endcapped) for effective sample cleanup
- Centrifuge tubes for convenient handling and processing
- Enhances retention of non-polar compounds in QuEChERS extractions
- Pre-measured sorbent ensures consistent and reliable results
- Streamlines laboratory workflow and reduces preparation time
Especificaciones
Especificaciones
| For Use With (Application) | QuEChERS |
| Product Line | Quechers |
| Sorbent | Sodium Sulfate |
| Type | QuEChERS Sample Prep |
| Volume (Metric) | 50 mL |
| Method | QuEChERS sample preparation |
| Sample Size | 500 mg per sample |
| Sample Type | All |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.