missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ 3 Quantitation Starter Kit

Product Code. 15397463
Change view
Click to view available options
Type:
Quantitation Starter Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15397463 Quantitation Starter Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15397463 Supplier Invitrogen™ Supplier No. Q33217

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Starter kit for the Qubit 3.0 Fluorometer that includes Qubit dsDNA BR, dsDNA HS, RNA HS, and Protein Assay Kits

The Qubit 3.0 Fluorometer accurately measures DNA, RNA, and protein using the highly sensitive fluorescence-based Qubit quantitation assays. The fluorescent dyes used in the these assays emit signals only when bound to specific target molecules, even at low concentrations, thus minimizing the effects of contaminants, including degraded DNA or RNA. The integrated design of the instrument and assays results in quantitation that is far more sensitive than UV absorbance, making this system ideal for quantitation of precious samples (samples that are rare, difficult to purify, or expensive to either obtain or prepare) or samples for delicate applications (samples for downstream assays that are extremely sensitive to sample conditions).

Qubit 3.0 Fluorometer Features

  • Powerful dual-core processor quickly and accurately quantifies DNA, RNA, and protein in less than 5 seconds per sample
  • Uses samples as little as 1μL up to 20μL
  • Stores up to 1000 sample results
  • Large, 5.7-inch, state-of-the-art color touch screen for easy workflow navigation
  • Ability to personalize Qubit fluorometer with the frequently run assays, add new assays, or even create custom assays with the MyQubit software and web tool
  • Small footprint saves bench space
  • Language options include English, French, Spanish, German, simplified Chinese and Japanese
  • Android-based operating system

Measurement of DNA, RNA, and protein using Qubit quantitation assays; Direct measurement of the fluorescence of samples; Evaluation of Ion Sphere™ Particle quality using the Ion Sphere Quality Control Kit prior to performing a sequencing run on the Ion PGM™ Sequencer

Certifications

Electromagnetic CE certification

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 Qubit 3 Fluorometer, 4 Qubit assays, and 500 Qubit assay tubes.
Store at room temperature. Reagents: dessicate and protect from light.
Sample Volume 1 to 20 μL
Includes Fluorometer, Assay Tubes, Assay Kits
Holds 1 Assay Tube per Read
Excitation Wavelength 430 to 495 nm Blue LED, 600 to 645 nm Red LED
For Use With (Equipment) Qubit Fluorometer
Product Line Qubit
Type Quantitation Starter Kit
Display Type 5.7 in. Capacitive Touchscreen
For Use With (Application) DNA Quantification, RNA Quantification, Protein Quantification
Memory 1000 Samples
Shipping Condition Room Temperature
Dimensions (L x W x H) 10 x 5.4 x 2.2 in. (25 x 13.6 x 5.5 cm)
Weight (Metric) 743.0 g
Voltage 100/240 VAC
Mostrar más Mostrar menos

For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.