missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Qubit™ 3.0 NGS Starter Kit

Product Code. 15324614
Change view
Click to view available options
Dynamic Range:
Five orders of magnitude
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dynamic Range unitSize
15324614 Five orders of magnitude Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15324614 Supplier Invitrogen™ Supplier No. Q33218

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Starter kit for next-generation sequencing with the Qubit 3.0 Fluorometer

The Qubit 3.0 Fluorometer accurately measures DNA, RNA, and protein using the highly sensitive fluorescence-based Qubit quantitation assays. The fluorescent dyes used in the these assays emit signals only when bound to specific target molecules, even at low concentrations, thus minimizing the effects of contaminants, including degraded DNA or RNA. The integrated design of the instrument and assays results in quantitation that is far more sensitive than UV absorbance, making this system ideal for quantitation of precious samples (samples that are rare, difficult to purify, or expensive to either obtain or prepare) or samples for delicate applications (samples for downstream assays that are extremely sensitive to sample conditions).

Qubit 3.0 Fluorometer Features

  • Powerful dual-core processor quickly and accurately quantifies DNA, RNA, and protein in less than 5 seconds per sample
  • Uses samples as little as 1μL up to 20μL
  • Stores up to 1000 sample results
  • Large, 5.7-inch, state-of-the-art color touch screen for easy workflow navigation
  • Ability to personalize Qubit fluorometer with the frequently run assays, add new assays, or even create custom assays with the MyQubit software and web tool
  • Small footprint saves bench space
  • Language options include English, French, Spanish, German, simplified Chinese and Japanese
  • Android-based operating system

Certifications

Electromagnetic CE certification

TRUSTED_SUSTAINABILITY

Specifications

Detector Type Photodiodes, measurement capability from 300 to 1000nm
Includes Qubit 3.0 Fluorometer (Mfr. No. Q33216); Qubit Assay Tubes, Pk. of 500 (Mfr. No. Q32856); and Qubit dsDNA HS Assay Kit, 500 assays (Mfr. No. Q32854)
Excitation Wavelength Blue: 430 to 495 nm, Red: 600 to 645 nm
Display Type Touch Screen
Dynamic Range Five orders of magnitude
Emission Wavelength Green: 510-580nm, Red: 665-720nm
Lamp Blue LED (max. ∽470nm), Red LED (max. ∽635nm)
Memory Instrument: 1000 samples, USB: 4GB
Processing Time ≤5 sec./sample
Dimensions (L x W x H) 10 × 5.4 × 2.2 in. (25 × 13.6 × 5.5cm)
Weight (English) 1.6 lb.
Weight (Metric) 743 g
Voltage 100/240 VAC
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.