Learn More
Skytec™ Quartz Safety Gloves
Product Code. 30274620
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274620
Brand: Skytec™ SKY443
Only null left
Add to basket
Add to basket
Ideal for precise operations requiring excellent dexterity and for preventing product contamination. A very supple, stretchable and comfortable mechanical barrier that adapts to every movement.
  • Lightweight with a Breathable PU Palm
  • Abrasion and tear resistant with a secure grip
  • Thin coating facilitates movement
  • Protects from oils and grease
  • Elasticated wrist provides a secure
  • Lint-free
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Abrasion 4,Puncture 2, EN 388:2016 4131X, EN 388:2016 + A1:2018 4131X
Construction Seamless Knit
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type General Purpose Gloves
Size Large
Length (Metric) 260 mm