missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ Absolute Q™ Digital PCR Starter Kit

Product Code. 17444683
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17444683 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17444683 Supplier Applied Biosystems™ Supplier No. A52732

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The QuantStudio Absolute Q Digital PCR Starter Kit is designed to confirm the performance of the QuantStudio Absolute Q Digital PCR System either at the time of system installation or as part of future, user-defined system verification procedures.

The QuantStudio Absolute Q Digital PCR Starter Kit is designed to confirm the performance of the QuantStudio Absolute Q Digital PCR System either at the time of system installation or as part of future, user-defined system verification procedures. The Starter Kit may also be used for training users on the QuantStudio Absolute Q Digital PCR System workflow.

The Starter Kit contains Absolute Q DNA Digital PCR Master Mix (5X) (Cat. No. A52490), Absolute Q dPCR Starter Assay, and CEPH DNA Control, providing a complete set of starter reagents and assay for use with the QuantStudio Absolute Q MAP16 Plate Kit (Cat. No. A52865).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 - Absolute Q™ DNA Digital PCR Master Mix 1- Absolute Q™ dPCR Starter Assay (20 x conc) 1- Control DNA CEPH 1347-02. Store at -15°C to -25°C.
Format Tube
PCR Method Digital PCR
Technique Digital PCR
For Use With (Equipment) QuantStudio™ Absolute Q™ Digital PCR System
Product Line Absolute Q
Quantity 1 Kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostics procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.