Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ AutoRun dPCR Suite, single configuration upgrade
Product Code. 30054825 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 upgrade
Unit Size:
Each
Product Code. Quantity Unit Size
30054825 1 upgrade Each
1 options
Product Code. 30054825 Supplier Applied Biosystems™ Supplier No. A57610
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
The Applied Biosystems QuantStudio Absolute Q AutoRun Digital PCR Suite is an integrated system solution that scales precise nucleic acid quantification and offers flexible instrument configurations and intelligent automation support for up to 72 hours of hands-free operation.
Upgrade your existing Applied Biosystems QuantStudio Absolute Q Digital PCR System to the QuantStudio AutoRun Digital PCR Suite, Single Configuration, which is an integrated system solution that scales precise nucleic acid quantification and offers intelligent automation to support hands-free operation.
With the QuantStudio Absolute Q AutoRun Digital PCR Suite you can:
  • Increase QuantStudio Absolute Q dPCR system utilization with multi-day unattended operation capabilities
  • Reclaim valuable operator time with continuous plate processing that does not require operator input
  • Simplify coordination and prioritization for multi-user systems
  • Enhance productivity with an intuitive software for setup, monitoring, and analysis, including powerful multi-plate studies
The QuantStudio Absolute Q AutoRun Digital PCR Suite upgrade adds the Spinnaker Microplate Mover to your existing QuantStudio Absolute Q dPCR System to manage loading and unloading of plates for an automated workflow. The easy-to-use Momentum Workflow Scheduler Software lets you manage AutoRun dPCR Suite multi-plate batches using the Spinnaker Microplate Mover.
Plate storage and handling
Two pre-configured plate storage and handling options are available: random access hotels and sequential access stacks.
Random access hotels offer additional flexibility for multi-user operation by accommodating the addition of plates as well as plate processing prioritization changes while a batch is already in process.
Powerful and intuitive software
QuantStudio Absolute Q dPCR software is designed to complement our intuitive workflow to save time and effort. Parameters such as plate format, optical channels, and thermal conditions can be easily modified to generate data quickly.
The clean and powerful data analysis interface provides a range of options for processing data. Whether visualizing multiplexed data or calculating concentrations across replicates, results can be exported for downstream analysis or compiled into an easy-to-read report in just a few clicks. Additionally, the software scales to your needs with multi-plate and multi-batch analysis capabilities.
Included security, auditing, and e-signature (SAE) software features support compliance with 21CFR part 11 for regulated environments.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Absolute Q
Voltage 100/240 VAC
Product Type Sample Loader
For Use With (Application) Digital PCR
Quantity 1 upgrade
Calibration Type Positional Teaching