Learn More
Applied Biosystems™ QuantStudio™ Absolute Q™ AutoRun Digital PCR Suite, single to dual configuration upgrade
Product Code. 30056277 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 upgrade
Unit Size:
Each
Product Code. Quantity unitSize
30056277 1 upgrade Each
1 options
Product Code. 30056277 Supplier Applied Biosystems™ Supplier No. A57612
Upgrade your existing QuantStudio Absolute Q AutoRun Digital PCR Suite, Single Configuration to the QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, an integrated system solution that scales precise nucleic acid quantification and offers intuitive automation to help support productivity and accelerate progress.
Upgrade your existing QuantStudio Absolute Q AutoRun Digital PCR Suite, Single Configuration, to the QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, an integrated system solution that scales precise nucleic acid quantification and offers intuitive automation to help support productivity and accelerate progress.
The QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, consists of two QuantStudio Absolute Q Digital PCR systems integrated with the Spinnaker Microplate Mover via the easy-to-use Momentum Workflow Scheduler Software to manage loading and unloading of plates for an automated workflow. By harnessing the power of two digital PCR systems in your automated workflow, you can double your throughput while fortifying the resilience of your operations.
This upgrade includes a location bracket, a nest adapter, necessary cables, and an updated Thermo Scientific Momentum Workflow Software license to support integration of the second QuantStudio Absolute Q Digital PCR System.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Absolute Q™
Voltage 100/240 V
For Use With (Application) Digital PCR
Quantity 1 upgrade
Calibration Type Positional Teaching