missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 7 PRO 0.2mL, 96-well package

The Applied Biosystems QuantStudio 7 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Offers facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.

Brand:  Applied Biosystems™ A44293


Additional Details : Weight : 71.00000kg

 View more versions of this product

Product Code. 16217641

  • £71443.00 / Each
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Features of the QuantStudio 7 Pro Real-Time PCR System include:
  • Easy and convenient hands-free operation with facial authentication and voice commands 
  • Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID 
  • Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen 
  • Peace of mind with Smart Help; contact Technical Support directly from the touchscreen 
  • Reduced downtime with Smart Remote Support; show issues to Technical Support for speedy resolution 
  • Application versatility with easily changed blocks and four different block types 
  • Increased productivity with automation compatibility for 24/7 operation 
  • Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing 
  • Remotely monitor experiments using the Instrument Connect monitoring app 
  • Design and Analysis software compatible with PC and Mac operating systems 
  • Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • Provides exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation 
  • Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab
Specifications

Specifications

PC (Tower)
96
54.7 cm
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
4°C to 99.9°C
QuantStudio 7 PRO
0.2 mL (96-well)
CMOS Technology
38 kg
0.4°C
Touchscreen
33.8 cm
52.5 cm
6.5°C/s
Bright White LED
5/6 Targets
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
96-well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers