missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 7 Flex package

Delivers the proven reliability, sensitivity, and accuracy of the Applied Biosystems ViiA 7 System in a new industrial design. The QuantStudio 7 Flex System has been optimized to enable the broadest range of quantitative PCR applications, with additional dyes, formats, and automation options.

Brand:  Applied Biosystems™ A27188


Additional Details : Weight : 14.23000kg

 View more versions of this product

Product Code. 15731178

  • £76625.00 / Each
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
  • The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
Specifications

Specifications

PC (Tower)
384
54.7 cm
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
4°C to 99.9°C
384-well: 3.5°C/s
Optiflex System Lamp
38 kg
0.25°C
Touchscreen
33.8 cm
52.5 cm
QuantStudio 7 Flex
3.0°C/s (384-well)
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
384-well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers