missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
- The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
Specifications
Specifications
| Content And Storage | PC (Laptop) |
| Thermal Uniformity | 0.25°C |
| Capacity | 96 |
| Display Type | Touchscreen |
| Height (Metric) | 54.7 cm |
| Width (Metric) | 33.8 cm |
| Warranty | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
| Depth (Metric) | 52.5 cm |
| Thermal Range | 4°C to 99.9°C |
| Description | QuantStudio 7 Flex |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
