missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio 7 Flex package
Shop All Applied Biosystems Products
Click to view available options
Content And Storage:
PC (Laptop)
PC (Tower)
Block Configurations:
384-well
96-well
96-well FAST
TaqMan Array Card
Warranty:
1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year)
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
Unit Size:
Each
Description
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
- The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
Specifications
Specifications
| Content And Storage | PC (Laptop) |
| Thermal Uniformity | 0.25°C |
| Capacity | 96 |
| Display Type | Touchscreen |
| Height (Metric) | 54.7 cm |
| Width (Metric) | 33.8 cm |
| Warranty | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
| Depth (Metric) | 52.5 cm |
| Thermal Range | 4°C to 99.9°C |
| Description | QuantStudio 7 Flex |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
