missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio 7 Flex package

Product Code. 15711168 Shop All Applied Biosystems Products
Click to view available options
Content And Storage:
PC (Laptop)
PC (Tower)
Block Configurations:
384-well
96-well
96-well FAST
TaqMan Array Card
Warranty:
1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year)
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
missing translation for 'unitSize'
Each
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Product Code. 15711168

missing translation for 'mfr': Applied Biosystems™ A27174

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Delivers the proven reliability, sensitivity, and accuracy of the Applied Biosystems ViiA 7 System in a new industrial design. The QuantStudio 7 Flex System has been optimized to enable the broadest range of quantitative PCR applications, with additional dyes, formats, and automation options.

  • Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
  • The QuantStudio™ 6 Flex system accommodates theversatility of 4 interchangeable blocks
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage PC (Laptop)
Thermal Uniformity 0.25°C
Capacity 96
Display Type Touchscreen
Height (Metric) 54.7 cm
Width (Metric) 33.8 cm
Warranty 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year)
Depth (Metric) 52.5 cm
Thermal Range 4°C to 99.9°C
Description QuantStudio 7 Flex
Max. Heating Rate (Thermal Block) 96-well: 3.9°C/s
Max. Cooling Rate 3.6°C/s (96-well)
Excitation Optiflex System Lamp
Volume (Metric) Thermal Block Sample 0.2 mL (96-well)
Pre-Calibrated Dyes FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
Weight 38 kg
Block Configurations 96-well
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.