Learn More
Applied Biosystems™ QuantStudio 6 PRO 0.2mL, 96-well package
The Applied Biosystems QuantStudio 6 Pro Real-Time PCR System adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint. Offers facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface.
£61243.00 - £62143.00
Specifications
Display Type | Touchscreen |
---|---|
Warranty | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
Product Code | Brand | Content And Storage | Warranty | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Content And Storage | Warranty | Price | Quantity & Availability | ||||
16267631
|
Applied Biosystems™
A44288 |
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£61243.00
Each |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16287631
|
Applied Biosystems™
A44290 |
PC (Laptop) | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£62143.00
Each |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16207641
|
Applied Biosystems™
A44292 |
PC (Tower) | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£62143.00
Each |
Estimated Shipment: 26-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Applied Biosystems QuantStudio 6 PRO Real-Time PCR System has all of the features of the QS6 FLEX system wth added smart features to deliver a smart, innovative and lab-changing workflow experience, including:
- Facial authentication; Hands-free login and access to your personal profile
- Hands-free operation; Perform basic operations using voice commands
- Radio-frequency identification (RFID) smart plate workflow; 2 embedded RFID readers to detect and decode Applied Biosystems™ TaqMan™ Smart Plates
- Smart Help; For faster, more efficient, and more effective resolution of instrument and application issues
- Smart Remote Support; A real-time audio/video collaboration tool and an advanced remote desktop support tool that reduces instrument downtime from days to minute
- Exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation.
- Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
Specifications
Touchscreen | |
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |