missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio 6 Flex package
Facilitates real-time PCR based applications with an upgradable block selection. With a simplified workflow, intuitive software, and touch-screen interface, the QuantStudio™ 6 Flex system offers exceptional reproducibility at a price you can afford.
£63016.00 - £82828.00
Specifications
Thermal Uniformity | 0.25°C |
---|---|
Display Type | Touchscreen |
Height (Metric) | 54.7 cm |
Width (Metric) | 33.8 cm |
Depth (Metric) | 52.5 cm |
Product Code | Brand | Content And Storage | Block Configurations | Warranty | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Content And Storage | Block Configurations | Warranty | Price | Quantity & Availability | |||
15781168
|
Applied Biosystems™
A27181 |
PC (Laptop) | 96-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£65616.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15761158
|
Applied Biosystems™
A27167 |
PC (Laptop) | 96-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£79028.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15761168
|
Applied Biosystems™
A27179 |
PC (Laptop) | 96-well FAST | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£63016.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15771188
|
Applied Biosystems™
A27961 |
PC (Laptop) | 384-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£67816.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15791188
|
Applied Biosystems™
A27963 |
PC (Laptop) | 384-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£81228.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15771168
|
Applied Biosystems™
A27180 |
PC (Tower) | 96-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£65516.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15731158
|
Applied Biosystems™
A27164 |
PC (Tower) | 96-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£78928.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15751168
|
Applied Biosystems™
A27178 |
PC (Tower) | 96-well FAST | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£63016.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15741158
|
Applied Biosystems™
A27165 |
PC (Tower) | 96-well FAST | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£76428.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15751188
|
Applied Biosystems™
A27959 |
PC (Tower) | 384-well | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
£69416.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15781188
|
Applied Biosystems™
A27962 |
PC (Tower) | 384-well | 1 Year Manufacturer's + 3 Year Field Service repair (with 1 planned maintenance/Year) |
£82828.00
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
- Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
- Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
- The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block
- The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change
Specifications
0.25°C | |
54.7 cm | |
52.5 cm | |
QuantStudio 6 Flex | |
38 kg |
Touchscreen | |
33.8 cm | |
4°C to 99.9°C | |
Excitation 455 to 672 nm, Emission 505 to 723 nm |