missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye), 96-well, 0.1mL

Product Code. 17927931
This item is not returnable. View return policy
Product Code. 17927931 Supplier Thermo Scientific™ Supplier No. A45218
  • ONLINE EXCLUSIVE PRICE
    £127.00
    £123.48 / Pack of 1
    Save £3.52
    2% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calibration plate for establishing the pure dye spectra value for the Cy™5.5 dye on Applied Biosystems™ QuantStudio™ 5 96-well 0.1-mL real-time PCR systems.

This plate establishes the pure dye spectra value for the Cy™5.5 dye on Applied Biosystems™ QuantStudio™ 5 96-well 0.1-mL real-time PCR systems. The plate facilitates calibration of the 6th OptiFlex™ channel on the QuantStudio™ 5 system, enabling highly multiplexed PCR assays to be used. Please refer to the instrument user guide for full instructions on usage.

TRUSTED_SUSTAINABILITY

Specifications

Calibrated Dye Cy™5.5
For Use With (Equipment) Applied Biosystems™ QuantStudio™ 5 96-well 0.1-mL real-time PCR systems
Detection Method Primer-probe
Capacity 1 x 96-well Plate, 0.1 mL
Product Type Spectral Calibration Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Description QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye)
Quantity 1 Plate
GC-Rich PCR Performance High
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.