missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye), 96-well, 0.1mL
Description
This plate establishes the pure dye spectra value for the Cy™5.5 dye on Applied Biosystems™ QuantStudio™ 5 96-well 0.1-mL real-time PCR systems. The plate facilitates calibration of the 6th OptiFlex™ channel on the QuantStudio™ 5 system, enabling highly multiplexed PCR assays to be used. Please refer to the instrument user guide for full instructions on usage.
Specifications
Specifications
| Calibrated Dye | Cy™5.5 |
| For Use With (Equipment) | Applied Biosystems™ QuantStudio™ 5 96-well 0.1-mL real-time PCR systems |
| Detection Method | Primer-probe |
| Capacity | 1 x 96-well Plate, 0.1 mL |
| Product Type | Spectral Calibration Plate |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Dry Ice |
| Description | QuantStudio™ 5 Spectral Calibration Plate (Cy™5.5 dye) |
| Quantity | 1 Plate |
| GC-Rich PCR Performance | High |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction