Learn More
Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System
Beskrivelse
Carry out rapid food protection testing with the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System. The affordable and easy-to-use system is ideal for users who need superior performance, maximum dye versatility, and security options in a real time PCR system.
The QuantStudio™5 Food Safety System is a high performance benchtop platform combining ease-of-use and system connectivity. Benefit from the flexibility of a 6-channel, 96-well, cloud-enabled open platform suitable for running our full range of PCR solutions for food safety, authenticity, and quality testing.
- Convenient 96-well Format: minimizes the effort required to perform sample setup while also allowing the use of tube strips.
- Obtain performance and results you can trust – OptiFlex technology and white LED optics enable powerful and sensitive data collection which improves well-to-well and instrument-to-instrument accuracy. Detect differences in target quantity as small as 1.5 – fold in singleplex reactions, and obtain 10 logs of linear dynamic range.
- Save valuable time- Veriflex block technology provides 3 or 6 independent temperature zones for flexibility to run multiple experiments simultaneously. Fast thermal cycling is also available, enabling results in less than 30 minutes.
- Designed for interactivity: sensitive, interactive touch screen provides stand-alone capabilities, PIN-protected user accounts, and dye calibration/RNases P functionality.
- Efficient: facilitates processing of multiple assays in the same run.
- Simple Intuitive Software: enables rapid, straightforward PCR set-up, automatic data interpretation, and traceability of results. Latest in qPCR technology enabling cloud benefits, compatible with RapidFinder™ Analysis software for running validated SureTect™ PCR Assays, and Thermo Scientific™ RapidFinder™Salmonella Multiplex Assay.
- Maximize bench top space: compact instrument can be configured as standalone or with a computer to fit any laboratory needs.
- Dedicated Service and Support: with our highly experienced scientists and engineers who are devoted to offering rapid response best-in-class support. Extended service and support packages are available. From preventative maintenance to corrective services, from loan instrument to field application specialists, get dedicated, local support from the experts in real-time PCR. Factory Calibrated: minimal calibration recommended (every two years)
- Get started quickly: instrument is factory-calibrated for optical and thermal accuracy and delivered in a user-friendly package for quick installation and immediate use.
The QuantStudio 5 Food Safety System is a real-time PCR system that delivers high-speed cycling with a new, fast ramping Peltier-based thermal cycling block. Coupled with an Optical Fast 96-well plate or 8-well strips and a hot-start DNA polymerase, inclusive of a master mix, environmental master mix and/or SureTect™ PCR Assay the QuantStudio 5 Food Safety System delivers complete runs in as fast as 30 mins, depending on your application.
- Pre-calibrated dyes at installation: FAMTM/SYBR® Green, VIC®/JOETM/HEX/TET, ABY/NEDTM/TAMRATM/Cy3®, JUN, ROXTM/Texas Red™, MUSTANG PURPLE, Cy®5/LIZ™
Tekniske data
Tekniske data
| Content And Storage | QuantStudio 5, 96well 0.1ml Block Real Time PCR System, Desktop tower computer RapidFinder Analysis software |
| Display Type | Touchscreen |
| For Use With (Application) | QuantStudio 5 Food Safety Real Time PCR System |
| Description | PCR System w/ Desktop and RapidFinder Analysis Software |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.