Learn More
Description
The QuantStudio™ 12K System’s intuitive software, easy touch screen setup, automation integration, effortless block exchange, and easy-to- use OpenArray™ components are designed to let you get started right away and reduce hands on time. Plan for the future and save time, money, and space in your lab with the only all-in-one qPCR instrument enabling a broad range of applications with maximum throughput.
- Includes five interchangeable blocks: The QuantStudio™ 12K blocks accomodate 96-well, FAST 96-well, 384-well, TaqMan™ array cards, and OpenArray™ plates, for effortless scaling from 1 to 12,000 data points in a single run. Increase throughput as you dive deeper into your research, using just one instrument. As you begin your project, start with 96-well format experiments and then scale up to a higher throughput to get the statistically significant answers you need. Additionally, you can easily validate and screen your targets of interest with a selected panel of genes using novel formats such as the TaqMan™ array cards and OpenArray™ plates.
- Compatible with a full range of TaqMan™ assays: The QuantStudio™ 12K System is optimized to enable clear, clean data for all TaqMan™ assays that might be used across a broad range of applications, including gene expression, microRNA and small noncoding RNAs, SNP genotyping, protein analysis, protein thermal shift, high resolution melt, copy number variation, and pathogen detection. • Provides a seamless switch from qPCR to digital PCR: Increase the precision and sensitivity of your nucleic acid experiment by switching from qPCR to digital PCR mode using QuantStudio™ Digital PCR kits and DigitalSuite™ Software with the QuantStudio™ OpenArray™ block. • Delivers peace of mind: Leave early on a Friday and monitor runs from home, knowing that the system can safely store over 100 runs. • Enhanced OptiFlex™ System: Building upon Applied Biosystems’ proprietary technology, the Enhanced OptiFlex™ System provides enhanced fluorescence detection for the accurate and sensitive data analysis necessary to collect 12,000 data points in a single run.
- Unprecedented coverage and sample throughput: The OpenArray™ format accelerates genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.
- Simple OpenArray™ workflow: QuantStudio™ Open Array™ plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the Accufill™ System. Easily load four arrays into the QuantStudio™ 12K instrument to achieve over 12,000 data points. Integrated sample tracking software lets you easily track your samples throughout the course of your project.
- Integrated analysis and quality systems: New intuitive touch-screen interface and integrated calendar feature allows for scheduling time of use on the instrument. Additional analysis software such as DigitalSuite™ v1.0, ExpressionSuite v1.0, and Genotyper v1.1 is available to help you analyze hundreds of targets and samples and achieve the statistical power you need.
- High Resolution Melt (HRM): Using the built-in MeltDoctor™ protocols and calibrations, you can spend less time optimizing the run and more time customizing your results. Define the number of variants you want to see by quickly changing analysis settings.
Specifications
Specifications
| Content And Storage | PC (Laptop) |
| Thermal Uniformity | 0.25°C |
| Capacity | 384 |
| Display Type | Touchscreen |
| Height (Metric) | 73.8 cm |
| Width (Metric) | 50.5 cm |
| Warranty | 1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance) |
| Depth (Metric) | 67.2 cm |
| Thermal Range | 4°C to 99.9°C |
| Max. Ramp Rate | 3.0°C/s |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
