missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Digital PCR Starter Kit
Shop All Applied Biosystems Products
Click to view available options
:
1 kit
Description
A variety of starter kits are available to introduce you to the various applications of the QuantStudio™12K Flex OpenArray™ sample block. Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments. The Digital PCR starter kit contains 2 OpenArray™ Digital PCR plates. Note that the QuantStudio™12K Flex Software (included with the QuantStudio™ 12K Flex System) offers example experiment files for each application.
The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates.
The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates.
Specifications
Specifications
| Content And Storage | • 2 OpenArray™ Digital PCR Plates • 1 bottle TaqMan™ OpenArray™ Digital PCR Master Mix • 1 tube Human Male DNA • 1 tube TaqMan™ Assay, RNase P • 1 kit OpenArray™ Accessories |
| Detection Method | Primer-probe |
| Format | OpenArray Plate |
| PCR Method | Digital PCR |
| Polymerase | AmpliTaq Fast DNA Polymerase |
| For Use With (Equipment) | QuantStudio™ 12k Flex |
| Label or Dye | FAM |
| No. of Tests | 2 Tests |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | OpenArray™, TaqMan™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction