missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Nitrite 3000 Test Strip Kits

Product Code. 10127480 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
Yellow to Red
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10127480 Yellow to Red Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127480 Supplier Macherey-Nagel™ Supplier No. 91322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows for the quick and easy determination of nitrate and nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrate 3000 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Useful for checking cooling lubricants and drinking water
  • Rapid: provides reliable results within one minute
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator Yellow to Red
Format Strip
For Use With (Application) Determination of Nitrite 3000
Graduations 0, 100, 300, 600, 1000, 2000, 3000mg/L NO2-
Quantity 100 Pack
Storage Requirements 30°C
Test Time 30 sec.
Type Semi-qualitative Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.