missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
Specifications
| Color Indicator | Pink to Red |
| Format | Strip |
| For Use With (Application) | Determination of Aluminum |
| Graduations | 0, 5, 20, 50, 200, 500mg/L Al3+ |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 2 min. |
| Type | Semi-qualitative Test Strip |
Safety and Handling
Product Identifier
- QUANTOFIX Aluminium
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: potassium hydroxidesolution, acetic acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?