missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 973 for H/M/R BMP-2

Product Code. 30001890
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001890 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001890 Supplier R&D Systems™ Supplier No. QC49
  • ONLINE EXCLUSIVE PRICE
    £165.54
    £147.00 / Pack of 1
    Save £18.54
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog numbers DBP200, SBP200, PDBP200

Quantikine Immunoassay Control Set 973 is designed as a quantitative control for the determination of BMP-2 cytokine concentrations in biological fluids.

  • Application: Control
  • Species Reactivity: Human, Mouse, Rat
  • Content: Recombinant human cytokines at low, medium, and high concentrations in a buffered protein solution with preservatives
  • Format: Lyophilized
  • Storage: Store unopened product at 2°C to 8°C
  • Stability: Stable for at least 6 months from date of receipt when stored at 2°C to 8°C
  • Intended Use: For research use only
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes BMP-2
Sample Type Biological Fluids
Type Immunoassay Control
Gene ID (Entrez) 29373
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Human, Mouse, Rat
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.