missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 867 Human TREM-1
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | TREM1 |
| Gene ID (Entrez) | 54210 |
| Intended Use | Control |
| Quantity | 1 Each |
| Source | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction