missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 834 for Human Fibronectin
Description
These controls are designed for use with R and D Systems™Quantikine™ kits to provide quantitative determination of cytokine concentrations in biological fluids.
- Form: Lyophilized
- Composition: Buffered protein solution containing recombinant human cytokines at low, medium and high concentrations
- Storage: 2°C to 8°C
- Stability: At least 6 months at 2°C to 8°C from date of receipt; for reconstituted controls, users should evaluate stability at <-20°C or discard after use
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detectable Analytes | Fibronectin |
| Type | Immunoassay Control |
| Gene ID (Entrez) | 2335 |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
| Source | Human |
| Form | Lyophilized |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?