missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 695 for Human SHBG

Product Code. 30001885
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001885 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001885 Supplier R&D Systems™ Supplier No. QC158B
  • ONLINE EXCLUSIVE PRICE
    £165.54
    £147.00 / Pack of 1
    Save £18.54
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as quantitative controls for the determination of cytokine concentrations in biological fluids

  • Prepared in diluted porcine serum with preservatives
  • Contains recombinant human cytokines at low, medium and high concentrations
  • Supplied lyophilized
  • Storage: Unreconstituted Controls should be stored at 2° to 8°C and are stable for at least 6 months from date of receipt. Depending on the analyte of interest, reconstituted controls may be stable when stored at <-20°C. Users should evaluate the frozen stability of the controls in their application or discard after use.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 189°C
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Detectable Analytes Sex Hormone-binding Globulin (SHBG)
Sample Type Biological Fluids
Type ELISA Control
Gene ID (Entrez) 6462
Intended Use Control
Lyophilized Yes
Melting Point -40°C
Product Line R&D Systems
Quantity 1 Each
Source Human
Stability Unreconstituted Controls are stable for at least 6 months from date of receipt. Depending on the analyte of interest, reconstituted controls may be stable when stored at <-20°C.
Form Liquid
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.