missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1078 for human PD-1
Click to view available options
Quantity:
Ea.
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | ELISA, Control |
| Quantity | Ea. |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction