missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, blood samples

Product Code. 31942694
Change view
Click to view available options
Quantity:
10 Plates
2 Plates
5 x 10 Plates
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31942694 5 x 10 Plates Each
32044639 2 Plates Pack of 2
32045382 10 Plates Pack of 10
32044491 5 x 10 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31942694 Supplier Invitrogen™ Supplier No. QS0112
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample

What you get
The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.

TRUSTED_SUSTAINABILITY

Specifica

Content And Storage • 5 x 50 mL Lysis Mixture, store at 15°C to 30°C
• 5 x 3.25 mL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type Blood
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 5 x 10 Plates
Shipping Condition Blue Ice
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.