missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Lysis Mixture

Product Code. 31942078
Change view
Click to view available options
Quantity:
12 mL
36 mL
120 mL
Unit Size:
120mL
12mL
36mL
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31942078 120 mL 120mL
32044367 12 mL 12mL
32044727 36 mL 36mL
32045505 120 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31942078 Supplier Invitrogen™ Supplier No. QP0524
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays.

QuantiGene Lysis Mixture is used to lyse cells from cell culture or whole blood, in order to release target RNA to be quantified with QuantiGene assays. QuantiGene Lysis Mixture is one of two components in the QuantiGene Sample Processing kits for cells and whole blood, and is intended to supplement these kits when additional volume is required.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Multi-Gene Expression
Product Line QuantiGene
Product Type Cell Lysis Buffer
Quantity 120 mL
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.