Learn More
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus epidermidis ATCC™ 12228™
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Brand: Thermo Scientific™ R4716500
660.25 GBP valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Code : 65
Additional Details : Weight : 0.40000kg Transport : UN number : 3373 Chem class : 6.2
Description
Thermo Scientific™ Quanti-Cult Plus™ Staphylococcus epidermidis ATCC™ 12228™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
- No serial dilution required
- Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
- 10 tests/reconstituted vial; 100 tests/kit
- Ready for rehydration
- For use in quality control procedures
Includes:
10 preserved organism vials and 10 rehydration vials.
Specifications
12228TM* | |
Quanti-Cult Plus | |
Cocci | |
100 Test/Kit | |
FDA strain PCI 1200 |
Staphylococcus epidermidis ATCC™ 12228™ | |
Quality Control Organism | |
Quantitative QC | |
100 Tests/Kit |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.