missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Quanti-Cult Plus™ Escherichia coli ATCC™ 8739™

Product Code. 12976037
Change view
Click to view available options
Quantity:
100 Tests/Kit
Unit Size:
10 tests
Product Code. Quantity unitSize
12976037 100 Tests/Kit 10 tests
1 options
This item is not returnable. View return policy

Product Code. 12976037

Brand: Thermo Scientific™ R4717085

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.

Thermo Scientific™ Quanti-Cult Plus™ Escherichia coli ATCC™ 8739™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.

  • No serial dilution required
  • Delivers less-than100 colony forming units (CFUs) in a single 0.1mL inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures

Includes:

10 preserved organism vials and 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

ATCC® 8739TM*
Description Escherichia coli ATCC™ 8739™
Format Quanti-Cult Plus
Product Type Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 100 Test/Kit
Quantity 100 Tests/Kit

*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.